Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CoA Synthase Monoclonal Antibody (OTI3D5)
GREENER_CHOICE

Catalog No. PIMA525579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525579 Supplier Invitrogen™ Supplier No. MA525579
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CoA Synthase Monoclonal Antibody for Western Blot, Flow

Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase and dephospho-CoA kinase, in prokaryotes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CoA Synthase
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3D5
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COASY
Gene Accession No. Q13057
Gene Alias 1300003G02Rik; bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; CoA synthase; Coasy; Coenzyme A synthase; Dephospho-CoA kinase; Dephospho-CoA pyrophosphorylase; Dephosphocoenzyme A kinase; DPCK; DPCOAK; NBIA6; NBP; nucleotide binding protein; Pantetheine-phosphate adenylyltransferase; Phosphopantetheine adenylyltransferase; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2; PPAT; PSEC0106; Ukr1
Gene Symbols COASY
Host Species Mouse
Immunogen Full length human recombinant protein of COASY produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80347
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.