Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CoA Synthase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554626
Description
Immunogen sequence: RVFGNKKQLK ILTDIMWPII AKLAREEMDR AVAEGKRVCV IDAAVLLEAG WQNLVHEVWT AVIPETEAVR RIVERDGLSE AAAQSRLQSQ MSGQQLVEQS HVVLSTLWEP HITQRQVEKA WALLQKRIPK THQ Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase and dephospho-CoA kinase, in prokaryotes.
Specifications
CoA Synthase | |
Polyclonal | |
Unconjugated | |
COASY | |
1300003G02Rik; bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; CoA synthase; Coasy; Coenzyme A synthase; Dephospho-CoA kinase; Dephospho-CoA pyrophosphorylase; Dephosphocoenzyme A kinase; DPCK; DPCOAK; NBIA6; NBP; nucleotide binding protein; Pantetheine-phosphate adenylyltransferase; Phosphopantetheine adenylyltransferase; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2; PPAT; PSEC0106; Ukr1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
287711, 71743, 80347 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13057, Q9DBL7 | |
COASY | |
Recombinant protein corresponding to Human CoA Synthase. Recombinant protein control fragment (Product #RP-93171). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction