Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CoA Synthase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117880
This item is not returnable.
View return policy
Description
CoA Synthase Polyclonal Antibody for Western Blot, IP
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase and dephospho-CoA kinase, in prokaryotes.
Specifications
CoA Synthase | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q13057 | |
COASY | |
E. coli-derived Human COASY fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
COASY | |
1300003G02Rik; bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; CoA synthase; Coasy; Coenzyme A synthase; Dephospho-CoA kinase; Dephospho-CoA pyrophosphorylase; Dephosphocoenzyme A kinase; DPCK; DPCOAK; NBIA6; NBP; nucleotide binding protein; Pantetheine-phosphate adenylyltransferase; Phosphopantetheine adenylyltransferase; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2; PPAT; PSEC0106; Ukr1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
80347 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction