Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546211 Supplier Invitrogen™ Supplier No. PA546211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KSEGLLYVHS SRGGPFQRWH LDEVFLELKD GQQIPVFKLS GENGDEVKKE Sequence homology: Human: 100%.

Component of the MITRAC (mitochondrial translation regulation assembly intermediate of cytochrome c oxidase complex) complex, that regulates cytochrome c oxidase assembly. MITRAC complexes regulate both translation of mitochondrial encoded components and assembly of nuclear-encoded components imported in mitochondrion. Required for assembly of mitochondrial respiratory chain complex I and complex IV. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene COA1
Gene Accession No. Q9GZY4
Gene Alias C7orf44; COA1; Cytochrome c oxidase assembly factor 1 homolog; cytochrome c oxidase assembly protein 1 homolog; mitochondrial translation regulation assembly intermediate of cytochrome c oxidase protein of 15 kDa; MITRAC15
Gene Symbols COA1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human COA1 (aa 97-146).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55744
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.