Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570263 Supplier Invitrogen™ Supplier No. PA570263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 79%; Rabbit: 83%; Rat: 86%.

Putative COX assembly factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COA4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene COA4
Gene Accession No. Q9NYJ1
Gene Alias 5330414O08Rik; AI115477; AW047259; CHCHD8; CMC3; Coa4; coiled-coil-helix-coiled-coil-helix domain containing 8; coiled-coil-helix-coiled-coil-helix domain-containing protein 8; cytochrome c oxidase assembly factor 4; cytochrome c oxidase assembly factor 4 homolog; cytochrome c oxidase assembly factor 4 homolog, mitochondrial; E2ig2; E2-induced gene 2 protein; RGD1562228
Gene Symbols COA4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human COA4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51287
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.