Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563480
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563480 Supplier Invitrogen™ Supplier No. PA563480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKYYEDKPQG GACAGLKEDL GACLLQSDCV VQEGKSPRQC LKEGYCNSLK YAFFECKRSV LDNRARFR Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen COA5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COA5
Gene Accession No. Q86WW8
Gene Alias 6330578E17Rik; C2orf64; CEMCOX3; COA5; Cytochrome c oxidase assembly factor 5; Pet191; protein C2orf64
Gene Symbols COA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COA5. Recombinant protein control fragment (Product #RP-99698).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493753
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.