Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COA6 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP306320AF405

Catalog No. NB135727


Only null left
Add to Cart

Description

Description

COA6 Polyclonal specifically detects COA6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

COA6
Polyclonal
Immunocytochemistry/Immunofluorescence
chromosome 1 open reading frame 31, cytochrome c oxidase assembly factor 6 homolog (S. cerevisiae), hypothetical protein LOC388753
Produced in rabbits immunized with E. coli-derived Human COA6 fragment.
0.1 mL
Primary
Human
Purified
Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
388753
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.