Learn More
Invitrogen™ COA7 Monoclonal Antibody (1G11-1C4)

Description
Peptide Sequence: MAGMVDFQDE EQVKSFLENM EVECNYHCYH EKDPDGCYRL VDYLEGIRKN FDEAAKVLKF NCEENQHSDS CYKLGAYYVT GKGGLTQDLK AAARCFLMAC EKPGKKSIAA CHNVGLLAHD GQVNEDGQPD LGKARDYYTR ACDGGYTSSC FNLSAMFLQG APGFPKDMDL ACKYSMKACD LGHIWACANA SRMYKLGDGV DKDEAKAEVL KNRAQQLHRE QQKGVQPLTF G.
Specifications
Specifications
| Antigen | COA7 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1G11-1C4 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | COA7 |
| Gene Accession No. | Q96BR5 |
| Gene Alias | 2010305A19Rik; AI451324; beta-lactamase hcp-like protein; C1orf163; COA7; Cytochrome c oxidase assembly factor 7; cytochrome c oxidase assembly factor 7 (putative); D4Ertd796e; hcp beta-lactamase-like protein C1orf163; hcp beta-lactamase-like protein C1orf163 homolog; RESA1; respiratory chain assembly 1; respiratory chain assembly factor 1; RGD1305274; Sel1 repeat containing 1; sel1 repeat-containing protein 1; Selrc1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.