Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555779 Supplier Invitrogen™ Supplier No. PA555779
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GMVDFQDEEQ VKSFLENMEV ECNYHCYHEK DPDGCYRLVD YLEGIRKNFD EAAKVLKFNC EENQHSDSCY KLGAYYVTGK G Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

Required for assembly of mitochondrial respiratory chain complex I and complex IV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COA7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COA7
Gene Accession No. Q96BR5
Gene Alias 2010305A19Rik; AI451324; beta-lactamase hcp-like protein; C1orf163; COA7; Cytochrome c oxidase assembly factor 7; cytochrome c oxidase assembly factor 7 (putative); D4Ertd796e; hcp beta-lactamase-like protein C1orf163; hcp beta-lactamase-like protein C1orf163 homolog; RESA1; respiratory chain assembly 1; respiratory chain assembly factor 1; RGD1305274; Sel1 repeat containing 1; sel1 repeat-containing protein 1; Selrc1
Gene Symbols COA7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COA7. Recombinant protein control fragment (Product #RP-94364).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65260
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.