Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556300
Description
Immunogen sequence: PGFPKDMDLA CKYSMKACDL GHIWACANAS RMYKLGDGVD KDEAKAEVLK NRAQQLHKEQ QKGVQPLTFG Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Required for assembly of mitochondrial respiratory chain complex I and complex IV.
Specifications
COA7 | |
Polyclonal | |
Unconjugated | |
COA7 | |
2010305A19Rik; AI451324; beta-lactamase hcp-like protein; C1orf163; COA7; Cytochrome c oxidase assembly factor 7; cytochrome c oxidase assembly factor 7 (putative); D4Ertd796e; hcp beta-lactamase-like protein C1orf163; hcp beta-lactamase-like protein C1orf163 homolog; RESA1; respiratory chain assembly 1; respiratory chain assembly factor 1; RGD1305274; Sel1 repeat containing 1; sel1 repeat-containing protein 1; Selrc1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
298377, 65260, 69893 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q921H9, Q96BR5 | |
COA7 | |
Recombinant protein corresponding to Human COA7. Recombinant protein control fragment (Product #RP-94365). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction