Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor II/Thrombin Antibody (046), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290542JF646
Description
Coagulation Factor II/Thrombin Monoclonal antibody specifically detects Coagulation Factor II/Thrombin in Mouse samples. It is validated for ELISASpecifications
Coagulation Factor II/Thrombin | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
046 | |
ELISA | |
Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2 | |
Rabbit immunized with purified, recombinant Mouse Coagulation Factor II/Thrombin (Accession#: NP_034298.1, Met1-Gly618) | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Signal Transduction | |
2147 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction