Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor III/Tissue Factor Antibody (TF9-10H10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$207.00 - $560.50
Specifications
Antigen | Coagulation Factor III/Tissue Factor |
---|---|
Clone | TF9-10H10 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-10 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot, Immunofluorescence, Immunocytochemistry, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017696
![]() |
Novus Biologicals
NB100652210.025MG |
0.025 mg |
Each for $207.00
|
|
|||||
NB10065221
![]() |
Novus Biologicals
NB10065221 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Coagulation Factor III/Tissue Factor | |
1.0 mg/mL | |
Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot, Immunofluorescence, Immunocytochemistry, Flow Cytometry | |
Unconjugated | |
Mouse | |
Cancer | |
P13726 | |
2152 | |
Denatured Tissue factor isolated from human brain by the Factor VII affinity method. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
TF9-10H10 | |
Western Blot 1-10 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Primate, Human | |
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
F3 | |
IgG1 | |
Protein G purified | |
CD142, also known as Tissue Factor, is the membrane receptor for coagulation factors VII and VIIa and is the cell surface initiator of coagulation. It is the major molecule of this type and is criticial for controlling hemostasis, thrombosis and inflammation. NB100-65221 is specific for an epitope within the extracellular domain, epitope locus I. It recognizes both the reduced and native non-reduced human and primate tissue factors. It does not inhibit coagulation or neutralize factor VII binding to CD142. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title