Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor III/Tissue Factor Rabbit anti-Mouse, Clone: 34, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Coagulation Factor III/Tissue Factor |
---|---|
Clone | 34 |
Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Coagulation Factor III/Tissue Factor | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
Recombinant Mouse Coagulation Factor III/Tissue Factor Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
34 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cancer | |
0.2 um filtered solution in PBS | |
2152 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title