Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor VII Antibody (003), Alexa Fluor™ 700 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290368AF700
Description
Coagulation Factor VII Monoclonal antibody specifically detects Coagulation Factor VII in Mouse samples. It is validated for ELISA, ELISASpecifications
Coagulation Factor VII | |
Monoclonal | |
Alexa Fluor 700 | |
coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor VII (NP_034302.2; Met1-Leu446). | |
0.1 mL | |
Cardiovascular Biology | |
2155 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
003 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction