Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor X Antibody (3F2), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326595100UL
Description
Coagulation Factor X Monoclonal antibody specifically detects Coagulation Factor X in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Coagulation Factor X | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
2159 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
ELISA, Immunohistochemistry | |
3F2 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor | |
A synthesized peptide derived from Human Coagulation Factor X [UniProt P00742] | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction