Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor X Antibody (MM0192-10A10), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212198G
Description
Coagulation Factor X Monoclonal specifically detects Coagulation Factor X in Human samples. It is validated for Western Blot.Specifications
Coagulation Factor X | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
F10 | |
Human recombinant Coagulation Factor X | |
0.1 mL | |
2159 | |
Human | |
Purified |
Western Blot | |
MM0192-10A10 | |
Western Blot | |
coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor | |
Mouse | |
Protein G purified | |
Primary | |
Detects human Coagulation factor X and Xa | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction