Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coagulation Factor XI Antibody (MM0193-7C38), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212199F
Description
Coagulation Factor XI Monoclonal antibody specifically detects Coagulation Factor XI in Human samples. It is validated for Western Blot, NeutralizationSpecifications
| Coagulation Factor XI | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Neutralization | |
| MM0193-7C38 | |
| Western Blot, Block/Neutralize | |
| coagulation factor XI, EC 3.4.21, EC 3.4.21.27, FXIPlasma thromboplastin antecedent, MGC141891, PTA | |
| Human recombinant Coagulation Factor XI | |
| 0.1 mL | |
| Prostate Cancer | |
| 2160 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction