Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One High Altitude Coagulation Tubes with Sodium Citrate Solution
Click to view available options
Capacity (Metric):
1.8 mL
3.15 mL
Description
- For use in high altitude elevations above 5000 ft.
- 13 x 75 mm tubes filled with buffered tri-sodium citrate solution
- 3.2 % (0.109 mol/L) concentration
- Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- 18-month shelf life from date of manufacture
- Blue nonscrew safety pull cap can be removed with a gentle pull
Specifications
Specifications
| Product Type | High Altitude Sodium Citrate Blood Collection Tube |
| Additive | 3.2% Citrate Solution |
| Material | PET |
| Capacity (Metric) | 1.8 mL |
| Closure Color | Blue with White Ring |
| Tube Size | 13 x 75 mm |
| For Use With (Application) | High altitude elevations above 5000 ft. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction