Learn More
Eisco™ Coarse White Marble, Metamorphic Rock Specimen - Approx. 3 Inch
SCI_ED
Catalog No. S02921 Shop All Eisco Products
Change view
Click to view available options
Specimen Size:
3 in.
Catalog No. Specimen Size
S02921 3 in.
1 options
Catalog No. S02921 Supplier Eisco™ Supplier No. ESNG0057HAND
Only null left
Add to Cart
Add to Cart
For use in classroom activities to ascertain differences between metamorphic rocks and their provenance. EISCO Coarse White Marble Specimen (Metamorphic Rock) is formed from limestone. High-pressure, low-heat alteration leads to massive crystalline rock.
  • Marble is a metamorphic rock formed from limestone. The high-pressure, low-heat alteration leads to a massive crystalline rock that can form a variety of shapes. Marble is mined heavily for use in buildings and furniture, and has traditionally been used in architecture as well
  • Though much denser than limestone, marble will still effervesce in acid, making it useful in identification exercises
  • Each specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features
  • Our rock and mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
  • Samples are packaged in a transparent bag, labeled and barcoded for easy identification
  • While each specimen will vary slightly in shape, size and color, it will measure approximately 3 inch in length
  • Each specimen is selected and hand processed by an actual geologist, ensuring that each sample exhibits defining features
  • Rocks and minerals are sourced from a variety of locations around the world and are processed by hand in Victor, NY
Specifications
Product Type Metamorphic Rock
Specimen Size 3 in.