Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COASY Antibody (OTI3B3), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272224APC
Description
COASY Monoclonal specifically detects COASY in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| COASY | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| COASY | |
| Full length recombinant protein of human COASY (NP_079509) produced in HEK293T cells. | |
| 0.1 mL | |
| 80347.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI3B3 | |
| Flow Cytometry | |
| bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase, CoA synthaseUKR1, CoASY, Coenzyme A synthase, DPCK, FLJ35179, NBPbifunctional coenzyme A synthase, nucleotide binding protein, phosphopantetheine adenylyltransferase/ dephosphocoenzyme A kinase, POV-2, PPAT | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction