Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COASY Antibody (OTI3B3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272224H
Description
COASY Monoclonal specifically detects COASY in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
COASY | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
COASY | |
Full length recombinant protein of human COASY (NP_079509) produced in HEK293T cells. | |
0.1 mL | |
80347.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3B3 | |
Western Blot, Immunohistochemistry | |
bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase, CoA synthaseUKR1, CoASY, Coenzyme A synthase, DPCK, FLJ35179, NBPbifunctional coenzyme A synthase, nucleotide binding protein, phosphopantetheine adenylyltransferase/ dephosphocoenzyme A kinase, POV-2, PPAT | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction