Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COASY Rabbit anti-Human, Clone: 7K2O6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | COASY |
---|---|
Clone | 7K2O6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
COASY Monoclonal antibody specifically detects COASY in Human samples. It is validated for Western BlotSpecifications
COASY | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
80347 | |
IgG | |
Affinity purified |
7K2O6 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase, CoA synthaseUKR1, CoASY, Coenzyme A synthase, DPCK, FLJ35179, NBPbifunctional coenzyme A synthase, nucleotide binding protein, phosphopantetheine adenylyltransferase/ dephosphocoenzyme A kinase, POV-2, PPAT | |
A synthetic peptide corresponding to a sequence within amino acids 465-564 of human COASY (Q13057). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title