Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COASY Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | COASY |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8404
![]() ![]() |
Proteintech Group Inc
129911AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TAOK1 is a TAO type protein kinase. TAOK1 expression has been documented in rat brain. ESTs have been isolated from human normal adrenal, bone and breast and cancerous duodenum, cervix and germ cell.Specifications
COASY | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q13057, Q9DBL7 | |
287711, 71743, 80347 | |
COASY Fusion Protein Ag3651 | |
Primary | |
-20°C | |
COASY |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CoA synthase, COASY, Coenzyme A synthase, Dephosphocoenzyme A kinase, DPCK, DPCOAK, NBP, pOV 2, PPAT, PPAT Dephospho CoA kinase, UKR1 | |
Coasy | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title