Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coatomer Subunit Delta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP132377
Description
Coatomer Subunit Delta Polyclonal specifically detects Coatomer Subunit Delta in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
Coatomer Subunit Delta | |
Polyclonal | |
Unconjugated | |
0.1M Tris, 0.1M Glycine, and 20% Glycerol (pH 7.0) with 0.01% Thimerosal | |
Archain, archain 1, archain vesicle transport protein 1, coatomer delta subunit, coatomer protein complex, subunit delta, coatomer protein delta-COP, coatomer subunit delta, COPD, Delta-coat protein, delta-COP | |
Rabbit | |
57 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.76mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
P48444 | |
ARCN1 | |
Recombinant protein encompassing a sequence within the center region of human COPD. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
372 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction