Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) acetate, anhydrous, Co 32% min.

Catalog No. p-4781862
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2313814 25 g
AA2313822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2313814 Supplier Thermo Scientific Chemicals Supplier No. 02313814
Only null left
Add to Cart

CAS: 71-48-7 | C4H6CoO4 | 177.02 g/mol

Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-48-7
Molecular Formula C4H6CoO4
Molecular Weight (g/mol) 177.02
MDL Number MFCD00008689
InChI Key QAHREYKOYSIQPH-UHFFFAOYSA-L
Synonym cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
PubChem CID 6277
ChEBI CHEBI:85138
IUPAC Name cobalt(2+);diacetate
SMILES [Co++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 298°C (decomposition)
Color Pink to Purple
Physical Form Powder
Quantity 25 g
Assay Percent Range 32% min. as Co
Solubility Information Soluble in waterSoluble in water,alcohol,dilute acids and pentyl acetate(tetrahydrate).
Formula Weight 177.02
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cobalt(II) acetate, Anhydrous, Co 32% min.
Hazard Category H317-H334-H341-H350i-H360F
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 200-755-8
RTECSNumber AG3325000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.