Learn More
Cobalt(II) acetate tetrahydrate, 99.999% (metals basis)
Description
Cobalt(II) acetate tetrahydrate is used to prepare complexes for examination of the properties of metals with unusual coordination geometries. It is also used as a catalyst for oxidation and esterification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼140°C (in -4H2O) |
| Color | Red |
| Physical Form | Crystalline |
| Quantity | 2 g |
| Linear Formula | Co(OOCCH3)2·4H2O |
| UN Number | UN3077 |
| Beilstein | 3731397 |
| Merck Index | 14,2433 |
| Solubility Information | Soluble in water(380g/L). |
| Formula Weight | 249.08 (177.02 Anhydrous) |
| Show More |
Safety and Handling
| Hazard Category | H302-H317-H334-H335-H341-H350i-H360F |
| Hazard Statement | GHS H Statement H334-H350-H360-H341-H302-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Suspected of causing genetic defects. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 200-755-8 |
| RTECSNumber | AG3325000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.