Alfa Aesar™ Cobalt(II) benzoate

Manufacturer: Alfa Aesar™ 8738009

Quantity 10g

 View more versions of this product

Catalog No. AA8738009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 932-69-4
    Molecular Formula C14H10CoO4
    Linear Formula Co(OOCC6H5)2
    Formula Weight 301.16
    MDL Number MFCD00050769
    Chemical Name or Material Cobalt (II) benzoate

    Specifications

    Formula Weight 301.16
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    MDL Number MFCD00050769
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Odor Odorless
    TSCA TSCA
    Health Hazard 1 Warning
    Color Gray
    Melting Point 114° to 116°C
    Quantity 10g
    Molecular Formula C14H10CoO4
    Linear Formula Co(OOCC6H5)2
    CAS 932-69-4
    Sensitivity Hygroscopic
    Chemical Name or Material Cobalt (II) benzoate

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit