Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) chloride, 97%, anhydrous, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
7646-79-9
Cl2Co
129.84
MFCD00010938
cobalt ii chloride,cobaltchloride,cobalt chloride 0.1 m solution,cobalt ii chloride, anhydrous,cobalt ii-chloride,curator_000013,cobalt chloride anhydrous,wln: co g2,cobalt-ii-chloride
7646-79-9
Cl2Co
129.84
MFCD00010938
cobalt ii chloride,cobaltchloride,cobalt chloride 0.1 m solution,cobalt ii chloride, anhydrous,cobalt ii-chloride,curator_000013,cobalt chloride anhydrous,wln: co g2,cobalt-ii-chloride
Assay Percent Range 97%
Linear Formula CoCl2
Fieser 01,155; 06,127; 08,291; 10,101; 13,249; 14,99; 15,97
Merck Index 15, 2421
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Formula Weight 129.84
Physical Form Powder
Percent Purity 97%
Density 3.35
Iron (Fe) 500ppm max.
Lead (Pb) 100ppm max.
Nickel (Ni) 2000ppm max.
Packaging Glass Bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Sublimation Point 500 °C
Boiling Point 1049.0°C
Color Blue
Melting Point 735.0°C
Quantity 5g
Chemical Name or Material Cobalt(II) chloride, 97%
Show More Show Less

GHS H Statement
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage fertility.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cau

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF

GHS Signal Word: Danger

EINECSNumber : 231-589-4

RTECSNumber : GF9800000

TSCA : TSCA