Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent

Catalog No. ac423570050
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic Bottle

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 86.0°C
Color Lilac
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula CoCl2·6H2O
Fieser 01,155; 06,127; 11,479
Merck Index 15, 2421
Solubility Information Solubility in water: 970g/L (20°C)
Formula Weight 237.93
Percent Purity 98 to 102%
Grade ACS Reagent
Packaging Glass bottle
Vapor Pressure Negligible
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage fertility.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing genetic defects.
May cause cancer by inhalation.
May cause an allergic skin reaction.
May cause allergy or ast
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
RTECSNumber GG0200000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.