Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) chloride hexahydrate, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
Packaging:
Plastic bottle
7791-13-1
Cl2Co·6H2O
237.93
MFCD00149652
7791-13-1
Cl2Co·6H2O
237.93
MFCD00149652
Linear Formula CoCl2·6H2O
Fieser 01,155; 06,127; 11,479
Merck Index 15, 2421
Solubility Information Solubility in water: 970g/L (20°C)
Formula Weight 237.93
Physical Form Powder
Percent Purity 98.0 to 102.0 % (Complexometry)
Grade Analysis
Calcium Oxide 100ppm max.
Copper (Cu) 10ppm max.
Insoluble in Water 0.03% max.
Insoluble Matter 0.03 % max. (in water)
Iron (Fe) 50ppm max.
Lead (Pb) 10ppm max.
Magnesium (Mg) 20ppm max.
Magnesium Oxide 10ppm max.
Manganese (Mn) 10ppm max.
Nickel (Ni) 100ppm max.
Packaging Plastic bottle
Silicon Dioxide 20ppm max.
Solubility Solubility in water: 970g/L (20°C).
Sulfur (S) 50ppm max.
Color Purple to Red
Melting Point 86.0°C
Quantity 100g
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less

GHS H Statement
May damage fertility.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing genetic defects.
May cause cancer by inhalation.
May cause an allergic skin reaction.
May cause allergy or ast

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:

GHS Signal Word: Danger

RTECSNumber : GG0200000