Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis)

Catalog No. p-7048495
Click to view available options
Quantity:
10 g
50 g

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

Cobalt(II) chloride hexahydrate has been used to study apoptotic effects in HepG2 cells. Also reported as a stabilizer for reconstitution of the radiopharmaceutical technetium-99m exametazime. It has been used in studies to examine heat shock protein expression of inflamed skin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 87°C
Color Purple-Red
pH 4.9
Physical Form Crystalline Aggregates
Quantity 10 g
Linear Formula CoCl2·6H2O
UN Number UN3077
Merck Index 14,2437
Solubility Information Soluble in water,acetone,ether,and alcohols.
Formula Weight 237.93 (129.84 Anhydrous)
Percent Purity 99.998%
Grade Puratronic™
Odor Odorless
Sensitivity Hygroscopic
Density 1.924 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less
Hazard Category H302+H332-H317-H334-H341-H350i-H360F
Hazard Statement GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-589-4
RTECSNumber GG0200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.