Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) citrate dihydrate, 98%

Catalog No. AAB2408822
Click to view available options
Quantity:
25 g
100 g

CAS: 6424-15-3 | C12H20Co3O16 | 597.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6424-15-3
Molecular Formula C12H20Co3O16
Molecular Weight (g/mol) 597.076
MDL Number MFCD02684007
InChI Key SKMIIAIULKZNRQ-UHFFFAOYSA-N
Synonym cobalt ii citrate dihydrate
PubChem CID 131882668
IUPAC Name cobalt;2-hydroxypropane-1,2,3-tricarboxylic acid;dihydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.[Co].[Co].[Co]

Specifications

Melting Point ∼150°C (decomposition)
Quantity 100 g
UN Number UN3077
Formula Weight 591.02 (554.99 Anhydrous)
Percent Purity 98%
Chemical Name or Material Cobalt(II) citrate dihydrate
Hazard Category H302-H317-H334-H341-H350-H360
Hazard Statement GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 212-751-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.