Alfa Aesar™ Cobalt(II) hydroxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 1252422

Quantity 100g

 View more versions of this product

Catalog No. AA1252422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21041-93-0
    Linear Formula Co(OH)2
    Formula Weight 92.95
    MDL Number MFCD00016015
    Merck Index 14,2442
    Chemical Name or Material Cobalt(II) hydroxide

    Specifications

    Formula Weight 92.95
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P312-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H317
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Odor Odorless
    CAS 21041-93-0
    Melting Point 250°C (dec.)
    RTECS Number GG0904500
    Physical Form Powder
    Health Hazard 1 Warning
    Sensitivity Air Sensitive
    Linear Formula Co(OH)2
    Merck Index 14,2442
    Color Pink to purple
    Quantity 100g
    Solubility Information Soluble in acids, ammonia; Very slightly soluble in water
    TSCA TSCA
    MDL Number MFCD00016015
    Chemical Name or Material Cobalt(II) hydroxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H302-H317
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P312-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit