Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II,III) oxide, Thermo Scientific Chemicals

Catalog Number AAA1612118
Click to view available options
Quantity:
50 g
250 g
1000 g
1308-06-1
Co3O4
240.80
MFCD00010939
UBEWDCMIDFGDOO-UHFFFAOYSA-N
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
11651651
oxocobalt;oxo(oxocobaltiooxy)cobalt
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
1308-06-1
Co3O4
240.80
MFCD00010939
UBEWDCMIDFGDOO-UHFFFAOYSA-N
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
11651651
oxocobalt;oxo(oxocobaltiooxy)cobalt
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 900°C (decomposition)
Quantity 50 g
UN Number UN3077
Merck Index 14,2429
Solubility Information Insoluble in water.
Formula Weight 240.8
Density 6.11
Chemical Name or Material Cobalt(II,III) oxide
Hazard Category H334-H350i
Hazard Statement GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-157-2
RTECSNumber GG2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit