Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II,III) oxide, Puratronic™, 99.9985% (metals basis)

Catalog No. p-7048498
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1069509 10 g
AA1069518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1069509 Supplier Thermo Scientific Chemicals Supplier No. 01069509
Only null left
Add to Cart

CAS: 1308-06-1 | Co3O4 | 240.80 g/mol

It is used in artificial photosynthesis. It is used as a magnetic semiconductor. It is also employed in supercapacitors and lithium-ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-06-1
Molecular Formula Co3O4
Molecular Weight (g/mol) 240.80
MDL Number MFCD00010939
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
PubChem CID 11651651
IUPAC Name oxocobalt;oxo(oxocobaltiooxy)cobalt
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]

Specifications

Melting Point 900°C (decomposition)
Color Black
Physical Form Powder
Quantity 10 g
UN Number UN3077
Merck Index 14,2429
Solubility Information Insoluble in water.Soluble in acids and alkalies.
Formula Weight 240.8
Percent Purity 99.9985%
Grade Puratronic
Density 6.11 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II,III) oxide
Show More Show Less
Hazard Category H334-H350i
Hazard Statement GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-157-2
RTECSNumber GG2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.