Learn More
Cobalt(II,III) oxide, Puratronic™, 99.9985% (metals basis)
Description
It is used in artificial photosynthesis. It is used as a magnetic semiconductor. It is also employed in supercapacitors and lithium-ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 900°C (decomposition) |
| Color | Black |
| Physical Form | Powder |
| Quantity | 10 g |
| UN Number | UN3077 |
| Merck Index | 14,2429 |
| Solubility Information | Insoluble in water.Soluble in acids and alkalies. |
| Formula Weight | 240.8 |
| Percent Purity | 99.9985% |
| Grade | Puratronic™ |
| Show More |
Safety and Handling
| Hazard Category | H334-H350i |
| Hazard Statement | GHS H Statement H351-H317 Suspected of causing cancer. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 215-157-2 |
| RTECSNumber | GG2500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.