Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) iodide, anhydrous, 99.5% (metals basis)

Catalog No. p-7049816
Click to view available options
Quantity:
25 g
100 g

Moisture and humidity indicator, catalyst | CAS: 15238-00-3

Cobalt(II) iodide is used as a moisture and humidity indicator. It is also used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15238-00-3
MDL Number MFCD00016030
Synonym cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis

Specifications

Boiling Point 570°C (vac.)
Melting Point 515°C (vac.)
Physical Form Powder
Quantity 25 g
Merck Index 14,2443
Solubility Information Soluble in water
Formula Weight 312.74
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Assay (metals basis)
Chemical Name or Material Cobalt(II) iodide, Anhydrous
Show More Show Less
Hazard Category H317-H341-H350
Hazard Statement GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 239-283-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.