Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) isopropoxide

Catalog No. aa4281003
Click to view available options
Quantity:
1 g
5 g

CAS: 60406-95-3 | C6H14CoO2 | 177.109 g/mol

Cobalt(II) isopropoxide is used in display & imaging, catalysis & synthesis and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60406-95-3
Molecular Formula C6H14CoO2
Molecular Weight (g/mol) 177.109
MDL Number MFCD00799075
InChI Key VBXHMSMVQZJJLD-UHFFFAOYSA-N
Synonym cobalt ii isopropoxide, cobalt isopropoxide, acmc-20alem, cobalt ii diisopropoxide, cobalt 2+ dipropan-2-olate, 2-propanol, cobalt 2+ salt
PubChem CID 18517386
IUPAC Name cobalt(2+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].[Co+2]

Specifications

Packaging Ampule
Quantity 1 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 178.11
Physical Form Powder
Chemical Name or Material Cobalt (II) isopropoxide
Hazard Category H228-H315-H319-H335-H341
Hazard Statement GHS H Statement
H228-H341-H315-H319-H335
Flammable solid.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P240-P241-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.