Learn More
Cobalt(II) nitrate hexahydrate, Puratronic™, 99.999% (metals basis)
Description
It is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 55°C to 56°C |
Color | Red |
Physical Form | Wet Crystalline Aggregates |
Quantity | 50 g |
UN Number | UN1477 |
Merck Index | 14,2444 |
Solubility Information | Soluble in water,alcohol,acetone and ammonia. |
Formula Weight | 291.04 (182.94 Anhydrous) |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Show More |
Safety and Handling
Hazard Category | H272-H302+H332-H317-H318-H334-H341-H350i-H360F |
Hazard Statement | GHS H Statement H272-H334-H350-H360-H341-H302-H317 May intensify fire; oxidizer. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Suspected of causing genetic defects. Harmful if swallowed. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P210-P220-P221-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S. |
EINECSNumber | 233-402-1 |
RTECSNumber | QU7355500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.