Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) oxalate dihydrate, Reagent Grade

Catalog No. AA8775814
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

Stabilizer for HCN, temperature indicator | CAS: 5965-38-8 | C2H4CoO6 | 182.981 g/mol

Cobalt(II) oxalate dihydrate is used as a stabilizer for hydrogen cyanide and as a temperature indicator. It is used in the preparation of cobalt catalysts, and cobalt metal powder for powder-metallurgical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-38-8
Molecular Formula C2H4CoO6
Molecular Weight (g/mol) 182.98
MDL Number MFCD03414148
InChI Key MWHSMSAKVHVSAS-UHFFFAOYSA-L
Synonym cobaltousoxalate,cobaltoxalate
PubChem CID 131881712
IUPAC Name 1,3-dioxa-2-cobaltacyclopentane-4,5-dione dihydrate
SMILES O.O.O=C1O[Co]OC1=O

Specifications

Melting Point 229°C to 231°C (decomposition)
Density 3.021 g/mL
Odor Odorless
Quantity 25 g
UN Number UN3288
Merck Index 14,2445
Solubility Information Soluble in aqueous ammonia; Insoluble in water; Slightly soluble in acids.
Formula Weight 182.98 (146.95 Anhydrous)
Grade Reagent
Physical Form Powder
Chemical Name or Material Cobalt (II) oxalate dihydrate
Show More Show Less
Hazard Category H302+H312-H350
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 212-409-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.