Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cobalt(II) phosphate, anhydrous, 98%

Click to view available options
Quantity:
25g
100g
13455-36-2
Co3O8P2
366.739
MFCD00016028
ZBDSFTZNNQNSQM-UHFFFAOYSA-H
cobalt phosphate,cobaltous phosphate,cobalt ii phosphate,tricobalt bis orthophosphate,unii-0b5m38t47h,cobalt ii phosphate, anhydrous,phosphoric acid, cobalt 2+ salt 2:3,3co.2po4,cobalt ii phosphate octahydrate,phosphoric acid,cobalt 2+ salt 2:3
61615
cobalt(2+);diphosphate
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
13455-36-2
Co3O8P2
366.739
MFCD00016028
ZBDSFTZNNQNSQM-UHFFFAOYSA-H
cobalt phosphate,cobaltous phosphate,cobalt ii phosphate,tricobalt bis orthophosphate,unii-0b5m38t47h,cobalt ii phosphate, anhydrous,phosphoric acid, cobalt 2+ salt 2:3,3co.2po4,cobalt ii phosphate octahydrate,phosphoric acid,cobalt 2+ salt 2:3
61615
cobalt(2+);diphosphate
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
Formula Weight 366.74
Percent Purity 98%
Quantity 25g
Chemical Name or Material Cobalt(II) phosphate, anhydrous

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TB9279000

TSCA : TSCA