Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) stearate, Co 9-10%, Thermo Scientific™

Catalog No. AA4335218
Change view
Click to view available options
Quantity:
50 g
5 kg
20 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4335218 50 g
AA43352A7 5 kg
AA43352B8 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4335218 Supplier Thermo Scientific Chemicals Supplier No. 4335218

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-88-6
Molecular Formula C36H70CoO4
Molecular Weight (g/mol) 625.89
MDL Number MFCD00054343
InChI Key AMFIJXSMYBKJQV-UHFFFAOYSA-L
Synonym cobalt ii stearate, cobaltous stearate, cobalt stearate, stearic acid cobalt ii salt, octadecanoic acid, cobalt salt, cobalt distearate, cobalt 2+ dioctadecanoate, stearic acid cobalt salt, cobalt 2+ ; octadecanoate
PubChem CID 6451168
IUPAC Name cobalt(2+);octadecanoate
SMILES [Co++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

Color Purple
Density 1.71 g/mL
Linear Formula (C18H35O2)2Co
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 625.89
Concentration or Composition (by Analyte or Components) Co 9 to 10%
Physical Form Pellets
Chemical Name or Material Cobalt (II) stearate
Hazard Category H317-H334-H350
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 213-694-7
RTECSNumber RG1212000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.