Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers

Catalog No. AC216270010
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum

CAS: 25134-21-8 | C10H10O3 | 178.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25134-21-8
MDL Number MFCD00167036
InChI Key KNRCVAANTQNTPT-UHFFFAOYNA-N
Synonym Methyl nadic anhydride, Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride

Specifications

Color Yellow
Density 1.2300g/mL
Boiling Point 275.0°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C10H10O3
Refractive Index 1.5060 to 1.5080
Quantity 1 kg
Beilstein 17, 461
Specific Gravity 1.23
Molecular Weight (g/mol) 178.19
Formula Weight 178.18
Percent Purity 95%
Physical Form Oily Liquid
Chemical Name or Material Methyl-5-norbornene-2, 3-dicarboxylic anhydride, Mixture of isomers
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.