Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) thiocyanate, 98+%

Catalog No. p-7027023
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 3017-60-5 | C2H2CoN2S2 | 177.105 g/mol

Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) thiocyanate is best known for Scott test, which is a proven screening test for the presence of cocaine.

Solubility
Soluble in water.

Notes
Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3017-60-5
Molecular Formula C2H2CoN2S2
Molecular Weight (g/mol) 177.105
MDL Number MFCD00010937
InChI Key IQHZUTZWNWRODJ-UHFFFAOYSA-N
PubChem CID 131877393
IUPAC Name cobalt;thiocyanic acid
SMILES C(#N)S.C(#N)S.[Co]

Specifications

Odor Odorless
Linear Formula Co(SCN)2
Quantity 10 g
UN Number UN3077
Solubility Information Soluble in water.
Formula Weight 175.1
Percent Purity ≥98%
Chemical Name or Material Cobalt(II) thiocyanate
Hazard Category H302+H312+H332-H350
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 221-156-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.