Learn More
Cobalt(III) 2,4-pentanedionate, Thermo Scientific Chemicals
Description
Cobalt(III) 2,4-pentanedionate is used as a polymerization catalyst, vulcanizing agent, pigment in resins and glass. It is also used for the preparation of monoporphyrinates. Further, it is used in kinetic studies of iron-sulfur proteins. In addition to this, it reacts with trimethylaluminum to prepare Me(Sub>3Al/Co(acac)3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 211°C (decomposition) |
Odor | Light |
Quantity | 100 g |
Sensitivity | Hygroscopic; Moisture sensitive |
Solubility Information | Soluble in methanol and halogenated solvents. Slightly soluble in acetone,aromatic solvents and aliphatic solvents. Insoluble in water. |
Formula Weight | 356.26 |
Physical Form | Powder |
Chemical Name or Material | Cobalt(III) 2,4-pentanedionate |
Safety and Handling
Hazard Category | H302-H317-H334-H350 |
Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 244-527-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.