Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cobalt(ous) Chloride, Hexahydrate
SCI_ED

Catalog No. S25851 Shop All Fisherbrand Products
Click to view available options
Concentration or Composition (by Analyte or Components):
0.1M
1M

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7791-13-1, 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt (II) Chloride, Cobalt Muriate Hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Pink-Purple
Quantity 500 mL
Assay Percent Range <75%w/v
Linear Formula CoCl2·6H2O
Formula Weight 237.95
Concentration or Composition (by Analyte or Components) 0.1M
Chemical Name or Material Colbalt Chloride Solution
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.