Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt silicide, 99% (metals basis)

Catalog No. p-4824684
Click to view available options
Quantity:
25 g
100 g

CAS: 12017-12-8 | CoSi2 | 115.10 g/mol

Cobalt silicide is widely used in chemical research. It is also used for the production and characterization of very-large-scale integration (VLSI) applications in semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12017-12-8
Molecular Formula CoSi2
Molecular Weight (g/mol) 115.10
MDL Number MFCD00016021
InChI Key QKWVWIGBHQUEHX-UHFFFAOYSA-N

Specifications

Melting Point 1277°C
Color Gray
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 115.11
Odor Odorless
Density 5.3
Chemical Name or Material Cobalt silicide
Hazard Category H315-H317-H319-H334-H335-H350
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c
EINECSNumber 234-616-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.