Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Sulfate, 7-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5390739 Shop All Avantor Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
PubChem CID 61444
IUPAC Name λ²-cobalt(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystals
Quantity 2.5 kg
Formula Weight 281.1
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.03kg
Chemical Name or Material Cobalt Sulfate, 7-Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.