Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt titanium oxide, 99.8% (metals basis excluding Ni), Ni <0.15%

Catalog No. AA4167018
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4167018 50 g
AA4167030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4167018 Supplier Thermo Scientific Chemicals Supplier No. 04167018
Add to Cart

CAS: 12017-01-5

Cobalt titanium oxide shows magnetic properties at room temperature and used in electronics and as spintronic devices. It is used to improve the gas sensing ability of hydrogen sulfide by hydrothermal method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12017-01-5
MDL Number MFCD00016022
Synonym Cobalt titanate

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 154.83
Percent Purity 99.8%
Assay (metals basis excluding Ni), Ni <0.15%
Chemical Name or Material Cobalt titanium oxide
Hazard Category H315-H317-H319-H334-H335-H350
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c
EINECSNumber 234-615-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.