Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobaltous Chloride CS, USP/EP Red Primary Solution, Ricca Chemical
SDP

Catalog No. 221032 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle

1 mL = 59.5 mg CoCl2.6H2O

Chemical Identifiers

CAS 7791-13-1, 7647-01-0, 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Red
Equivalence 1 mL = 59.5 mg CoCl2·6H2O
Grade EP/USP/Colorimetric Solution
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Cobaltous Chloride, Red Primary Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.